Corning® Pipette Tips, Universal, Corning
Catalog #: 29442-682
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Volume:1 - 200 µl
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Tip Color:Natural
- Packed:Rack Packed
- Sterile:-
- Supplier No.:4863
Specifications
About this item
Corning® universal fit tips are designed to provide a reliable fit with all major brand pipettors.
Graduation rings on all tips provide volume reference.
- Beveled orifice helps ensure accurate fluid delivery
- All tips are certified RNase-/DNase-free and non pyrogenic
- Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design
- Bulk packed tips are non sterile and very economical
Corning® universal fit tips are designed to provide a reliable fit with all major brand pipettors.
1 to 200 µl universal fit tips are graduated at the 10 µl, 50 µl, and 100 µl volumes.
Customers Who Bought This Also Bought