Corning® 96-Well Polypropylene Tubes, Corning
Catalog #: 29442-614
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Capacity:1.2 ml
- Sterility:Sterile
- Description:8-Cap Strips
- Packaging:12/Bag
- Supplier No.:4418
Specifications
About this item
PP tubes, PE tube caps.
- Composed of 96 PP tubes in a standard microplate format
- 1.2 ml tubes available individually or in strips of 8 tubes
- PE tube caps available in 8-cap strips
PP tubes, PE tube caps.
Packaging: Supplied bagged or racked.
Customers Who Bought This Also Bought