Corning® Costar® Reagent Reservoirs, for Transtar-96® Liquid Handling System, Corning
Catalog #: 29442-526
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Description:Costar® Transtar-96® Disposable Reservoir Liner, 12 channel, Sterile
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Cat. no.:29442-526
- Supplier No.:4877
Specifications
About this item
White PS, sterile, disposable.
Customers Who Bought This Also Bought