Corning® Permeable Supports, Polycarbonate (PC) Membrane, Corning
Catalog #: 29442-106
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Pore size:0.4 µm
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Cat. no.:29442-106
- Membrane Diameter:6.5 mm
- Cell Growth Area:0.33 cm²
- Supplier No.:3413
Specifications
About this item
75 mm Transwell® with 0.4 µm pore polycarbonate membrane insert, sterile.
- 10 µm thick translucent polycarbonate membrane
- Treated for optimal cell attachment
- Each insert is packaged in a 100 mm dish, 12 inserts per case
- Membrane must be stained for cell visibility
Produces a cell culture environment that closely resembles an in vivo state. An essential tool for the study of both anchorage-dependent and independent cell lines.
Customers Who Bought This Also Bought