Transwell® Clear Inserts, Polyester (PET) Membrane
Catalog #: 29442-084
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Culture area:0.33 cm²
- Membrane Material:Polyester
- Pore size:3.0 µm
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Cat. no.:29442-084
- Packaging:12/Plate, 4 Plates
- Membrane Diameter:6.5 mm
- Supplier No.:3472
Specifications
About this item
Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.
- w Produce a cell culture environment that closely resembles an in vivo state
- Co-culture cells with or without cell-to-cell contact
- Design a diversity of experiments using various pore sizes, membrane types, and coatings 10 µm transparent membrane
- Treated for optimal cell attachment
- Excellent visibility under phase contrast microscopy
- Supplied in multiple well plates
- Sterile
Allow polarized cells to carry out metabolic activities in a more natural manner because the cells feed both apically and basolaterally.
Customers Who Bought This Also Bought