Glass Microfiber Filter Paper Grade 151, Ahlstrom
Catalog #: 28496-176
Supplier: Ahlstrom
Specifications
- Ø:4.7 cm
- Particle retention:0.7 microns
- Flow rate:Slow
- Load capacity:Very High
- Cat. no.:28496-176
- Supplier No.:1510-0470
Specifications
About this item
Ahlstrom Grade 151 is a binder-free microfiber glass filter designed to retain extremely fine particles.
It is recommended for use in the Toxicity Characteristic Leaching Procedure (TCLP) as described in EPA Method 1311.
Customers Who Bought This Also Bought