Cell Culture Storage Bottles, Polystyrene, Corning
Catalog #: 28199-758
Supplier: Corning
Product available on GSA AdvantageĀ®
Specifications
- Capacity:250 ml (8.5 oz.)
- Graduation Interval:25 ml
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:RECs, ISO 14001 - Oneonta
- Cat. no.:28199-758
- Supplier No.:430281
Specifications
About this item
Disposable polystyrene bottles for storage of media, buffers and other aqueous solutions.
- Low profile, easy grip style has sides that facilitate handling
- Plug seal caps (45 mm) provide an airtight seal and help minimise the risk of contamination
- Bottles can be used with CorningĀ® vacuum filter systems
- Sterile, certified non-pyrogenic
Customers Who Bought This Also Bought