Nalgene® Filter Storage Bottle Receivers, Sterile, Thermo Scientific
Catalog #: 28199-214
Supplier: Thermo Fisher Scientific
Specifications
- Capacity:150 ml
- Cat. no.:28199-214
- Supplier No.:455-0150
Specifications
About this item
These receiver storage bottles feature an ergonomic design with tapered sides and grips for ease of handling.
- Integral no-drip lip facilitates pouring
- Easy-to-read graduations on two sides
- Non-cytotoxic, non-pyrogenic, radiation sterilized
Storage bottles have a 45 mm Ø closure and are designed for use with Nalgene® Rapid-Flow™ series bottle-top filters and filter units. A 1¹/₂ -turn threaded screw closure guarantees a leakproof seal and reduces pH shift of stored media. The storage bottles have at least 20% extra head space for the addition of serum or other additives to the filtrate.
Customers Who Bought This Also Bought