Order Entry
ContactUsLinkComponent
COMB FOR B2 SYSTEM
Combs for Owl™ EasyCast™ Mini Gel Electrophoresis System, Models B1, B2 and B3, Thermo Scientific
Catalog #: 27373-272
COMB FOR B2 SYSTEM
Combs for Owl™ EasyCast™ Mini Gel Electrophoresis System, Models B1, B2 and B3, Thermo Scientific
Catalog #: 27373-272
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Rapid-Load 25 (2X)-Tooth comb, 1.5 mm
  • Cat. no.:
    27373-272
  • For Use With:
    B2 and B3 Systems
  • Supplier No.:
    B2-RL-25D

Specifications

About this item

Use comb with your exisitng U.V

  • For use in Owl™ EasyCast™ Mini Gel Electrophoresis Systems
  • Cast smaller gels

For use in Owl™ EasyCast™ Mini Gel Electrophoresis Systems. Use comb with your exisitng U.V. Transmissible (UVT) gel tray and comb slots to cast smaller gels. Thickness: 1.5mm (1/16").

Transmissible (UVT) gel tray and comb slots to cast smaller gels.

Customers Who Bought This Also Bought