To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:B1A System with Buffer Exchange Ports
- Cat. no.:27372-228
- L×W×H:16 x 9.5 x 8 cm (65/16 x 33/4 x 35/32")
- Buffer Volume:400 ml
- Gel Dimensions:7 x 8 cm
- Supplier No.:B1A-BP
Specifications
About this item
Mini-Gel systems have a small footprint and can run 5–34 samples (for 400 and 600mL systems) or 8–48 samples (for 800mL systems) on one gel
Leak-resistant, gasketed ultraviolet transmission trays allow tape-free casting in the buffer chamber. The trays have two comb positions and allow DNA bands to be viewed on a transilluminator without removing the gel. Optional multiload UVT trays have 12 slots for multiple sample-running, and allow samples to be loaded directly from a 96-well plate into an agarose gel. This design is based on the fixed spacing required to accommodate multichannel pipets (at least eight channels) and 96-well plates.
Heavy-duty, one-piece combs eliminate the need for comb assembly. The SuperSafe™ safety lid eliminates the risk of accidental shock. System is supplied with a 0.91m (3') safety power cord, attached.
The buffer chamber features lane visualization strips, which are silkscreened on the platform to provide a clear view of sample wells. Lane markers and fluorescent rulers provide accurate fragment size determination of DNA when compared to known standards. Durable acrylic construction and research-quality components give the buffer chamber a long life. Platinum electrodes and gold-plated banana plugs are corrosion-protected, and electrode placement ensures a uniform electric field.