To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Model A3 System with Buffer Exchange Ports
- Cat. no.:27372-122
- L×W×H:53 x 32 x 10.5 cm (207/8 x 1219/32 x 41/8")
- Buffer Volume:4.5 L
- Gel Dimensions:23 x 40 cm
- Supplier No.:A3-1
Specifications
About this item
Horizontal systems for detailed RNA/DNA analysis deliver consistent, quality results
A1 Model Systems feature a small footprint, conserving bench space. A2 Model Systems are wider than the A1 models, and can run up to 42 samples per comb position. They accomplish gel runs in 1–4 hours. A1 and A2 models are available with or without buffer exchange ports. A3 Model System is an extra-long apparatus designed especially for screening a large number of samples. The UVT gel tray for this system has multiple comb slots for more flexibility in comb placement. Gel tray fits 25- and 50-tooth microtiter format combs for fast sample loading with a multichannel pipet. System is ideal for field reversal electrophoresis and RFLP analysis. Buffer exchange ports are standard with this model. It accomplishes gel runs in 1–6 hours.
Constructed of heavy-duty acrylic for heavy workloads. The UV transmissible (UVT) gel tray allows bands to be viewed on a transilluminator without the risk of tearing the gel during handling. Comb slots on the UVT tray are positioned so that up to four samples can be set to run equal lengths on the same gel. Leak-resistant gasketed end gates provide tape-free casting of agarose gels directly in the gel tray. Three-point leveling ensures flat, even gels. Platinum electrodes and gold-plated banana plugs are corrosion-protected. The SuperSafe™ safety lid eliminates the risk of accidental shock, and an attached 0.91m (3') long power cord enhances safety. Voltage requirements for all models is 20–250V.