Barnstead Replacement Cartridges, Thermo Scientific
Catalog #: 26303-234
Supplier: Thermo Fisher Scientific
Specifications
- Description:High Capacity Cartridge, Two-Bed Ion Exchange Resin
- For Use With:Bantam Deionizer Cartridge Systems, B-Pure™ Double/Single Holder and Harvey DI+ Cartridge Systems
- Capacity:1760 grains as CaCO₃
- Supplier No.:D0803
Specifications
About this item
Cartridges feature all polypropylene construction and are designed to fit certain NANOpure®, E-pure™, B-pure™, PCS, Bantam holder, or cooling loop systems.
Customers Who Bought This Also Bought