Accessories for Barnstead E-pure™ Water Purification Systems, Thermo Scientific
Catalog #: 26302-611
Supplier: Thermo Fisher Scientific
Specifications
- Description:Down Flow, individually packed cartridge, used in fourth position of quad style NANOpure and E-Pure systems only, ORGANICfree Catridges
- Cat. no.:26302-611
- Supplier No.:D5021