Barnstead Replacement Cartridges, Thermo Scientific
Catalog #: 26302-366
Supplier: THERMO FISHER SCIENTIFIC MX
Specifications
- Description:Combination Colloid/Organic Removal with Macroreticular, Activated Carbon Resin
- For Use With:NANOpure®, E-Pure™, B-Pure™
- Capacity:5000 gal.s
- Supplier No.:D0835
Specifications
About this item
Cartridges feature all polypropylene construction and are designed to fit certain NANOpure®, E-pure™, B-pure™, PCS, Bantam holder, or cooling loop systems.
Customers Who Bought This Also Bought