Barnstead Replacement Cartridges, Thermo Scientific
Catalog #: 26291-306
Supplier: Thermo Fisher Scientific
Specifications
- Description:Ultrapure and Organic Removal with Mixed-Bed, Activated Carbon Resin
- Cat. no.:26291-306
- For Use With:B-Pure™, Still Pretreatment
- Capacity:750 Grains as NaCl
- Supplier No.:D0832
Specifications
About this item
Cartridges feature all polypropylene construction and are designed to fit certain NANOpure®, E-pure™, B-pure™, PCS, Bantam holder, or cooling loop systems.
Customers Who Bought This Also Bought