Specifications
- Ø:100 mm
- Surface:
- Description:Dish, TC-treated surface
- Environmentally Preferable:Low Manufacturing Impact
- Height:20 mm
- Environmental certification:RECs, ISO 14001 - Oneonta
- Cat. no.:25382-428
- Packaging:20/Sleeve
- Growth Area:55 cm²
- Working Volume:11.0 - 16.5 ml
- Actual growth surface:80.5 mm
- Supplier No.:430167
Specifications
About this item
Optically clear PS. Available with a choice of treated surface. Corning® CellBIND® surface increases surface wetability for more even and consistent cell attachment. Ultra-Low Attachment dishes feature a covalently bound hydrogel layer that minimises cell attachment, protein absorption and cellular activation.
- With stacking beads to aid handling
- Supplied with vents to provide consistent gas exchange
- Sterilised by gamma radiation
- Certified non-pyrogenic
These disposable petri-style dishes are manufactured from optically clear virgin polystyrene. Dishes are equipped with stacking beads to aid in handling and feature vents to ensure consistent gas exchange. Sterilized by gamma irradiation.
The surface minimizes cell attachment, protein absorption, and enzyme activation. It is ideal for culturing stem cells, as it promotes ebryoid body formation from ES cells. Surfaces are stable, noncytotoxic, biologically inert, and nondegradable.