Spectra/Por® 7 Dialysis Membranes, MWCO 1000 to 50000
Catalog #: 25223-785
Supplier: Spectrum Laboratories
Small Business Enterprise
Specifications
- Ø:15 mm
- Cat. no.:25223-785
- MWCO:10 kD
- Flat Width:24 mm
- Supplier No.:132118
Specifications
About this item
Free of trace metal and sulfur for separations involving a possible sensitivity to such materials.
- Transparent and flexible
- Minimum sulphur and heavy metal contamination
Supplied wet in 0.1% sodium azide as a preserving agent. Just rinse in distilled water and condition in dialyzing buffer and membrane is ready for use.
Roll length: 5 m (16.4')
Customers Who Bought This Also Bought