Order Entry
ContactUsLinkComponent
Walter Crucible Holder
Walter Crucible Holder, PLASTICOID COMPANY INC
Catalog #: 24065-000
Supplier:  Plasticoid
Small Business Enterprise
Walter Crucible Holder
Walter Crucible Holder, PLASTICOID COMPANY INC
Catalog #: 24065-000
Supplier:  Plasticoid
Small Business Enterprise

Specifications

  • Description:
    Walter Crucible Holder
  • Cat. no.:
    24065-000
  • Supplier No.:
    VCHWGS

Specifications

About this item

Rubber crucible holder for use with Gooch crucibles, or similar crucibles, in the neck of a standard suction flask.

  • For use with crucibles 20 to 35 mL, and flasks 250 to 1000 mL
Delivery: Supplied with funnel.
Customers Who Bought This Also Bought