Order Entry
Canada
Orders LinkContactUsLinkComponent
VWR® Hand Tally Counter

VWR® Hand Tally Counter

VWR® hand tally counter, 85 g (3 oz.)
Catalog # 23609-102
Supplier:   VWR International
VWR® Hand Tally Counter
Catalog # 23609-102
Supplier:  VWR International
VWR® Hand Tally Counter
VWR® Hand Tally Counter

VWR® hand tally counter, 85 g (3 oz.)

Price
$114.50
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents

Compact unit to record industrial events, receive lab samples, take surveys, inventory items, total drops of liquid, count occurrences, and more. Made of metal for reliability, long life, and hard use. The rugged, heavy-duty chrome-finished case has a hinged finger loop for a sure grip.

  • Easy-to-use four-digit unit counts up to 9999
  • White numbers appear on a black background
  • A protective window seals out dust, dirt, and humidity
  • Finger pressure on the button produces an audible click which signals the count
  • Unit is zeroed with a twist of the reset knob

Dia. 4.5 cm (1.75"), width: 3 cm (11/8")

Specifications
  • Description:
    VWR® hand tally counter
  • Weight:
    85 g (3 oz.)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.