Order Entry
ContactUsLinkComponent
pH 4.01 Buffer (color coded pink)
Orion™ pH Buffer Bottles, Thermo Scientific
Catalog #: 23198-222
CAS Number:  
pH 4.01 Buffer (color coded pink)
Orion™ pH Buffer Bottles, Thermo Scientific
Catalog #: 23198-222
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Color:
    Pink
  • Description:
    pH 4.01 buffer
  • Size:
    5×60 ml
  • Supplier No.:
    910460

Specifications

About this item

Perform accurate and reproducible pH calibrations with Orion™ pH buffer bottles.

  • All bottles feature pH/temperature tables printed on the labels for accurate calibrations and NIST traceability
  • Manufactured under ISO 9000 quality standards
  • Available in pint (475 ml) bottles or portable five packs of 2 oz. (5×60 ml) bottles

The pH 4.01, 5.00, 7.00, and 10.01 buffers are color-coded for ease of use. All Orion™ pH buffers are traceable to NIST standard reference materials.

Customers Who Bought This Also Bought