ContactUsLinkComponent
Pyrex* Brand Cloning Cylinder
PYREX® Cloning Cylinder, Corning
Catalog #: 22877-254
Supplier:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®
Pyrex* Brand Cloning Cylinder
PYREX® Cloning Cylinder, Corning
Catalog #: 22877-254
Supplier:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Monterrey
  • Cat. no.:
    22877-254
  • O.D.×H:
    8 x 8 mm
  • Supplier No.:
    3166-8
Specifications
About this item
Cloning cylinders, made of borosilicate glass code 7740. Clone a single cell or group of cells by surrounding them with this open ended cloning cylinders. Dip the end of the cylinder into sterile silicone grease before pressing to the bottom of a culture dish to create an isolated well.
  • Reusable
  • Non sterile
  • Cylinders are autoclavable or can be dry heat sterilized
Customers Who Bought This Also Bought