Order Entry
ContactUsLinkComponent
VWR* Cheesecloth Wipers
VWR® Cheesecloth Wipers
Catalog #: 21910-107
Supplier:  VWR International
VWR* Cheesecloth Wipers
VWR® Cheesecloth Wipers
Catalog #: 21910-107
Supplier:  VWR International

Specifications

  • W×L:
    10.1 x 10.1 cm (4 x 4")
  • Cat. no.:
    21910-107
  • Packaging:
    Dispenser Box

Specifications

About this item

Use to wipe, clean, or dry cuvettes, delicate glassware, syringes, pipets, and pH electrodes.

Use to wipe, clean, or dry cuvettes, delicate glassware, syringes, pipets, and pH electrodes. They are ideal for filtering and cleaning.

  • Available in convenient dispenser box or on economical bolt
  • Inert to even harsh solvents
  • Mesh size: grade 10

Cheesecloth wipers are ideal for filtering and cleaning

Cheesecloth wipers are ideal for filtering and cleaning

Wipers meet cleanliness specifications of USP and Federal Specification CCC-G-101c.

Bleached, all-purpose cheesecloth wipers are brilliant white and certified 100% pure cotton of reagent-grade quality and purity. They feature a unique combination of cleanliness, softness, absorbency, and strength. Will not scratch the most sensitive surfaces. Absorb six times their weight and absorb both solvents and aqueous solutions.

Ordering information: Wipers 21910-107 can be unfolded to approximately 17.8×35.6 cm (7×14"). Wipers 21910-105 can be unfolded to approximately 45.7×91.4 cm (18×36").
Customers Who Bought This Also Bought