Specifications
- Blade No.:#11
- Supplier No.:4-411
Specifications
About this item
Scalpels feature plastic handles and razor-sharp, stainless steel blades for superior cutting and clean incisions
Size is clearly marked on each blade. Safety sleeve protects both user and blade. Individually packaged in sealed, peel-apart pouches. Five-year shelf life.