Feather® Sterile Surgical Blade, GF Health Products (Graham-Field)
Catalog #: 21899-530
Supplier: GF HEALTH PRODUCTS INC. TE
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Stailess Steel Blade
- Cat. no.:21899-530
- Blade Size:#11
- Supplier No.:2976#11
Specifications
About this item
Superior cutting quality, strength and corrosion resistance characterize these stainless steel, disposable blades.
- Individually wrapped and sealed in aluminum foil to maintain sterility, insure long shelf-life
- Blades available in several sizes
Customers Who Bought This Also Bought