VWR® Digital Counter with Keychain, Wrist Strap, and Carabiner
Catalog #: 21800-062
Supplier: VWR International
Specifications
- Description:Digital counter with keychain
- Cat. no.:21800-062
Specifications
About this item
Designed for ease-of-use and fail-safe counting, this lightweight digital counter features a water-resistant O-ring seal, five-digit display count, button click for count confirmation, and recessed reset to prevent accidental tampering.
- Easy-to-use design for fail-safe counting
- Water-resistant O-ring seal
- Recessed reset to prevent accidental tampering
Ordering information: Supplied with two-year constant-use battery and shock-resistant ABS plastic case.
Customers Who Bought This Also Bought