Order Entry
ContactUsLinkComponent
VWR* Q Clamp Holder
VWR® Q Clamp Holder
Catalog #: 21572-712
Supplier:  VWR International
Product available on GSA Advantage®
VWR* Q Clamp Holder
VWR® Q Clamp Holder
Catalog #: 21572-712
Supplier:  VWR International
Product available on GSA Advantage®

Specifications

  • Description:
    Q-Style Post Holder Clamp
  • Cat. no.:
    21572-712

Specifications

About this item

VWR® Q clamp holder for securing clamps to a rod.

  • Q clamp holder for use in custom applications
  • May be used with 21572-703 3-prong clamp head
  • Made of die cast nickel plated zinc
  • Grips support rods up to 0.5" (13 mm)
  • Threaded opening is 0.3" (7.9 mm)
Customers Who Bought This Also Bought