Glass Wool and Extractor, Ohio Valley
Catalog #: 21507-980
Supplier: Ohio Valley
Small Business Enterprise
Specifications
- Description:DMCS (Silanized) Treated Glass Wool, 50 g
- Cat. no.:21507-980
- Supplier No.:3352
Specifications
About this item
Borosilicate glass wool, either untreated or treated with a dimethyldichlorosilane solution to render the wool inert for use in gas chromatography.
Fiber Diameter: 8 micron
Glass wool extractor: Insert extractor in column, twist slightly to hook glass wool and pull.
Customers Who Bought This Also Bought