ContactUsLinkComponent
Sterilization Indicator Labels
Gamma Radiation Sterilization Indicator Labels, NAMSA®
Catalog #: 19872-038
Supplier:  NAMSA
Sterilization Indicator Labels
Gamma Radiation Sterilization Indicator Labels, NAMSA®
Catalog #: 19872-038
Supplier:  NAMSA
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Imprinted, 5000 Indicators per Roll
  • Supplier No.:
    CPI-R03
Specifications
About this item
Self-adhesive, nontoxic, chemical process indicators change from yellow to red upon exposure to gamma or electron beam radiation.
Labels are coated with a UV blocking agent for protection from exposure to UV light. A minimum of 15kGy (1.5Mrad) irradiation is recommended for sufficient endpoint color development. At greater dosage levels, the radiation indicators exhibit a darker shade of red. When exposed, imprinted indicators 19872-038 read “RED IS [gamma] EXPOSED.” Indicators are stable for at least 24 months from date of manufacture when they are stored between 15 to 30°C.
Certifications: Conform to AAMI ST60 and ISO 11140-1, Section 6 for Class 1 Process Indicators.
Customers Who Bought This Also Bought