Order Entry
ContactUsLinkComponent
VWR* Stackable Sharps Container Safety System
VWR® Sharps Container Systems, Multiple Colors and Styles
Catalog #: 19001-008
Supplier:  VWR International
VWR* Stackable Sharps Container Safety System
VWR® Sharps Container Systems, Multiple Colors and Styles
Catalog #: 19001-008
Supplier:  VWR International

Specifications

  • Capacity:
    2 gal.
  • Dimensions:
    25.4W×17.8D×28.6H cm (10W×7D×11.25"H)
  • Color:
    Red
  • Cat. no.:
    19001-008
  • Style:
    Stackable

Specifications

About this item

Sharps container systems provide solutions for the safe and effective containment and disposal of contaminated needles, syringes, and other sharps.

  • Puncture-resistant
  • Autoclave-safe and incinerator-safe
  • Transparent, locking lids
  • Needle removal ports

The stackable sharps containers are puncture-resistant and both autoclaves-safe and incinerator-safe. The tortuous path systems restrict hand entry. The extended neck sharps containers feature a funneled vertical drop and extended handles.

Customers Who Bought This Also Bought