BRAND PD-Tip™ Syringe Tips for Repeating Pipettors, BrandTech
Catalog #: 18900-380
Supplier: Brandtech
Specifications
- Cat. no.:18900-380
- For Use With:25 - 50 ml Tips, Non Sterile
- Supplier No.:702398
Specifications
About this item
BRAND PD-Tips™ fit common repeating pipettors using standard sized tips, including the BRAND HandyStep and the VWR® repeating pipettor.
The 1 and 10 ml tip sizes allow for more useful volumes when using 'half settings'. All 25 and 50 ml PD-Tip™ packs include one adapter, also available separately.
Tip size is automatically recognized by the HandyStep® electronic repeating pipettor (see 47727-662) and the Rainin® AutoRep E. BIO-CERT® sterile tips are individually wrapped and independently certified free of DNA, RNase, ATP, and endotoxins for molecular biology assays.
Ordering information: BRAND PD-Tips™ are supplied in recyclable packaging. BIO-CERT® sterile tips are individually wrapped. The Multi-Pack contains 20 each of seven different sizes of PD-Tip™ (0.5, 1, 1.25, 2.5, 5, 10, and 12.5 ml).
Customers Who Bought This Also Bought