Specifications
- Capacity:500 ml (16.9 oz.)
- Description:Ethyl Acetate
- Cat. no.:16649-925
- Cap Color:Green
Specifications
About this item
Easy-to-read, break-resistant dispensing bottles include DOT, CAS numbers, National Fire Protection Association (NFPA) codes and symbols, protective equipment symbols, and target organ information.
- Easy-to-read, low-density polyethylene dispensing bottles are break-resistant
- Intended for short-term dispensing use
- Include DOT, CAS numbers, NFPA codes and symbols, protective equipment symbols, and target organ information
Polypropylene nozzle assemblies are leakproof and color-coded to match bottle label.
Toluene bottle is solid red with white lettering and hazard information.Sodium Hypochlorite bottle is white LDPE for light-protection between 230 and 450 nm. Variety pack includes one each: acetone (red closure), ethanol (natural closure), isopropanol (yellow closure), methanol (green closure), toluene (red closure), and water (blue closure).
Caution: LDPE wash bottles are intended for short term dispensing use only and not for storage of acetone, dichloromethane, toluene, and 1,1,1-trichloroethane.