WHEATON® Black Plastic Caps for Media Bottles, DWK Life Sciences
Catalog #: 16299-248
Supplier: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Specifications
- Cap Size:15-415
- Cat. no.:16299-248
- Supplier No.:240463
Specifications
About this item
Solid-top lined caps are suitable for a variety of applications
Choose from rubber-, fluoropolymer resin-, polyethylene-, or metal foil-lined caps. Rubber-lined caps are for culture work. Fluoropolymer resin-lined caps are for maximum product safety. Both rubber- and fluoropolymer resin-lined caps are autoclavable. Polyethylene-lined caps are designed for distilled water, analytical standards, and reagents. Metal foil-lined caps are for organic solvents.
Customers Who Bought This Also Bought