Microlink® Screw Caps, WHEATON
Catalog #: 16299-210
Supplier: DWK Life Sciences
Small Business Enterprise
Specifications
- Cap Size:33-430
- Color:Black
- Description:PP/Silicone liners
- Material:PP
- Supplier No.:240106
Specifications
About this item
These screw caps are ideal for use with biological media, volatile solvents, corrosive chemicals and sensitive compounds.
- Liner is ultrasonically welded to the cap to eliminate the possibility of glue contamination and bond failure common with autoclaving
- Available in convenient lab pack quantities as well as larger case packs
- Low-compression-set silicone ideal for the storage of samples for prolonged period of time over wide temperature ranges
- PTFE or polypropylene on the exposed face of the liner imparts an inert,chemical-resistant surface
- Open top caps provide access with a syringe
- Autoclavable
Customers Who Bought This Also Bought