NALGENE® Polypropylene Screw Closures, Thermo Scientific
Catalog #: 16225-234
Supplier: Thermo Fisher Scientific
Specifications
- Cap Size:83B
- Cat. no.:16225-234
- Supplier No.:712160-0830
Specifications
About this item
Caps for large NALGENE® bottles and carboys
Note: Before autoclaving, set cap on top of container without engaging threads.
Caps 16225-204 and -234 are buttress-style. Autoclavable.
Customers Who Bought This Also Bought