Nalgene® Biohazardous Waste Containers, Thermo Scientific
Catalog #: 16211-138
Supplier: Thermo Fisher Scientific
Specifications
- Capacity:56.8 L (15 gal.)
- Dimensions:33O.D.x68.6H cm (13x27)"
- Cat. no.:16211-138
- Supplier No.:6370-0015
Specifications
About this item
Autoclavable polypropylene waste containers are ideal for secondary containment of biohazardous waste materials.
Containers comply with OSHA 29CFR 1910.1030 for use as protection against bloodborne pathogens.
- Convenient one-step sterilization, with no need to remove autoclavable bag
- Port opening in cover allows convenient one-handed disposal of materials, limiting exposure to infectious waste
- Holds standard-size autoclavable bags
- Red universal biohazard symbol is molded in English and Spanish
- Leakproof side and bottom construction
Autoclavable polypropylene waste containers are ideal for secondary containment of biohazardous waste materials.
Containers comply with OSHA 29CFR 1910.1030 for use as protection against bloodborne pathogens.
Customers Who Bought This Also Bought