Order Entry
ContactUsLinkComponent
Clear Standard Wide Mouths, Glass Only, Bulk Pack
Standard Wide Mouth Bottles, Clear, Qorpak
Catalog #: 16195-805
Supplier:  Qorpak
Small Business Enterprise
Clear Standard Wide Mouths, Glass Only, Bulk Pack
Standard Wide Mouth Bottles, Clear, Qorpak
Catalog #: 16195-805
Supplier:  Qorpak
Small Business Enterprise

Specifications

  • Cap Size:
    58-400
  • Capacity:
    237 ml (8 oz.)
  • Cat. no.:
    16195-805
  • Supplier No.:
    GLA-00871

Specifications

About this item

Clear, flint glass standard wide mouth bottles feature a wide opening making them the ideal general-purpose bottle.

Applications include pharmaceutical, chemical, environmental, food packaging and food storage.

Ordering information: Bottles 36319-155, 36319-195, 36319-199, 36319-219, 36319-225, and 89204-692 are supplied pre-cleaned for volatiles. Bottles 89176-848, 89176-850, 93001-560, 93001-562, 16054-206, 76487-214 and 10053-746 are supplied pre-cleaned and certified for semi-volatiles.
Customers Who Bought This Also Bought