Bottle Beakers® Medium Rounds, Wide Mouth, Qorpak
Catalog #: 16194-290
Supplier: Qorpak
Specifications
- Cap Size:48-400
- Capacity:120 ml (4 oz.)
- Cat. no.:16194-290
- Supplier No.:GLC-01504
Specifications
About this item
These flint glass bottles are excellent for use with biological and pathological specimens, but can also be used for storing industrial laboratory chemicals and reagents.
- Graduated in ml and ounces
- Mix, measure, and store in the same container
These flint glass bottles are excellent for use with biological and pathological specimens, but can also be used for storing industrial laboratory chemicals and reagents.
They are designed with a slight shoulder for easy pouring and handling.
Ordering information: Bottles 36319-325, 36319-327, 36319-329, 36319-330, and 36319-319 are supplied pre-cleaned for volatiles.
Customers Who Bought This Also Bought