Order Entry
ContactUsLinkComponent
Safety Coated Glass, Amber Wide Mouth Packer, Glass Only - Convenience Pack
Safety-Coated Packer Bottles, Wide Mouth, Amber, Qorpak
Catalog #: 16180-880
Supplier:  Qorpak
Small Business Enterprise
Safety-Coated Packer Bottles, Wide Mouth, Amber, Qorpak
Catalog #: 16180-880
Supplier:  Qorpak
Small Business Enterprise

Specifications

  • Cap Size:
    45-400
  • Cap Type:
    Bottles Only
  • Capacity:
    237 ml (8 oz.)
  • Cat. no.:
    16180-880
  • Supplier No.:
    GLA-00964

Specifications

About this item

Reduce breakage and lab mishaps with these safety coated amber wide mouth packer bottles.

  • Safety-coated packer bottles are made of glass

If the glass breaks, the plastisol coating contains the substance in the bottle long enough to allow for proper disposal.

Ordering information: Bottles 89096-730, 89096-732, 89096-734, 89096-736, 89096-738, and 89096-740 are supplied Ultra Cleaned. Bottles 93000-914, 93000-916, 93000-918, 93000-920, 89212-384, and 89212-386 are supplied vacuumed and ionized.
Customers Who Bought This Also Bought