Nalgene® Economy Bottles, High-Density Polyethylene, Narrow Mouth, Thermo Scientific
Catalog #: 16172-128
Supplier: Thermo Fisher Scientific
Specifications
- Cap Size:28-415
- Capacity:500 ml (16.9 oz.)
- Color:Natural
- Cat. no.:16172-128
- Supplier No.:2089-0016
Specifications
About this item
Economical, single-use, break-resistant bottles feature thinner walls and are more flexible than original Nalgene® bottles.
- Excellent chemical resistance to most acids, bases, and alcohols
- Guaranteed leakproof
- Available in natural or amber
- Narrow mouths are ideal for pouring
Ideal for short-term, general-purpose laboratory and field applications, such as collecting samples, or for storing working solutions like buffers, bench reagents, and diluted chemical aliquots. Natural bottles allow easy viewing of liquid level. Amber bottles reduce UV light transmission for protection of light-sensitive liquids.
Certifications: Amber bottles comply with U.S. Pharmacopoeia 23 requirements for maximum light transmission.
Delivery: Supplied with polypropylene screw closures.
Caution: Not recommended for hazardous and critical material containment.
Customers Who Bought This Also Bought