Snap-On Stoppers, WHEATON®
Catalog #: 16171-660
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Specifications
- Color:Red
- Description:Snap-On Stopper
- Material:Rubber
- Size:13×20 mm
- Cat. no.:16171-660
- Supplier No.:224100-180
Specifications
About this item
These snap-on stoppers provide maximum sealing for vials with a blow-back glass finish, which is a small ridge on the interior of the vials neck.
- Not recommended for vials with a straight-walled finish
The blow-back feature is typically found in vials from North American manufacturers.
The snap-on stopper locks into the blow-back ridge, providing a tight seal and keeping the stopper from backing out.
Customers Who Bought This Also Bought