Serum Bottles, Borosilicate Glass, WHEATON®, DWK Life Sciences
Catalog #: 16171-374
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Capacity:60 ml (2 oz.)
- Material:Clear Glass
- Cat. no.:16171-374
- Diameter ×H:41 x 91 mm
- Supplier No.:223746
Specifications
About this item
Serum bottles are ideal for long and short term sample storage, lyophilization, and as vaccine/injectable drug containers.
- Bottles fit most lyophilization applications
Clear bottles are manufactured from Wheaton 400 borosilicate molded glass, and conform to USP Type I requirements. Amber bottles are manufactured from Wheaton 500 borosilicate glass, and conform to USP Type I requirements. All bottles feature a 13Int.Ø×20Ext.Ø mm mouth opening, with the exception of 89422-806 which is 7Int.Ø×13Ext.Ø mm.
Ordering information: Bottles are packaged in shrink-wrapped, partitioned modules to reduce breakage.
Customers Who Bought This Also Bought