Order Entry
ContactUsLinkComponent
Leak-Resistant Bottle, Polypropylene, Narrow Mouth
Leak-Resistant Bottles, Polypropylene, Narrow Mouth, WHEATON®, DWK Life Sciences
Catalog #: 16155-169
Small Business Enterprise
Leak-Resistant Bottle, Polypropylene, Narrow Mouth
Leak-Resistant Bottles, Polypropylene, Narrow Mouth, WHEATON®, DWK Life Sciences
Catalog #: 16155-169
Small Business Enterprise

Specifications

  • Cap Size:
    28-410
  • Capacity:
    500 ml (16.9 oz.)
  • Dimensions:
    72 Dia. x 171 H, mm
  • Cat. no.:
    16155-169
  • Supplier No.:
    209169

Specifications

About this item

For general laboratory use. Polypropylene bottles feature a special pour lip to facilitate safe, clean dispensing.

  • Double-seal closure for leak-resistance
  • Autoclavable

Note: Before autoclaving, set cap on top of bottle without engaging threads.

Ordering information: Supplied in convenience-packed cartons with polypropylene caps attached.
Customers Who Bought This Also Bought