Order Entry
ContactUsLinkComponent
Leak-Resistant Bottle, High-Density Polyethylene, Wide Mouth
Leak-Resistant Bottles, High-Density Polyethylene, Wide Mouth, Wheaton®, DWK Life Sciences
Catalog #: 16153-513
Small Business Enterprise
Leak-Resistant Bottle, High-Density Polyethylene, Wide Mouth
Leak-Resistant Bottles, High-Density Polyethylene, Wide Mouth, Wheaton®, DWK Life Sciences
Catalog #: 16153-513
Small Business Enterprise

Specifications

  • Cap Size:
    63-415
  • Capacity:
    1 L (33.8 oz.)
  • Dimensions:
    91 Dia. x 199 H, mm
  • Cat. no.:
    16153-513
  • Supplier No.:
    209550

Specifications

About this item

Durable bottles are ideal for general laboratory use and field sample analysis.

  • Wide mouth design facilitates easy filling and pouring
  • Designed with a special pour lip to facilitate safe, clean dispensing
  • Double-seal closure for leak-resistance
Ordering information: Supplied in convenience-packed cartons with polypropylene caps attached.
Customers Who Bought This Also Bought