Order Entry
ContactUsLinkComponent
Clear Glass Boston Round Bottle
Boston Round Bottles, Clear, Narrow Mouth
Catalog #: 16151-614
Supplier:  Qorpak
Product available on GSA Advantage®
Small Business Enterprise
Clear Glass Boston Round Bottle
Boston Round Bottles, Clear, Narrow Mouth
Catalog #: 16151-614
Supplier:  Qorpak
Product available on GSA Advantage®
Small Business Enterprise

Specifications

  • Cap Size:
    24-400
  • Capacity:
    237 ml (8 oz.)
  • Cat. no.:
    16151-614
  • Supplier No.:
    GLA-00815

Specifications

About this item

Clear, flint glass Boston round bottles offer maximum visibility and sample integrity.

  • USP certified Type III (soda lime) glass

These general use bottles are perfect for liquids, product storage, and field or plant sampling projects. They feature a round body, rounded shoulders, and narrow screw neck opening. Environmentally sensitive bottles help to eliminate waste and help to ensure product integrity for long term storage.

Ordering information: Bottles 16152-203, 16152-209, 16152-211, 11311-644, 93001-592, 93001-626, 93001-628, 93001-636, and 93001-638 are supplied Ultra Cleaned. Bottles 36319-125, 36319-212, and 36319-245 are supplied Cleaned for Volatiles. Bottle 36319-140 is supplied Cleaned and Certified for Volatiles. Bottle 93001-654 is supplied Cleaned and Certified for Semi-Volatiles.
Customers Who Bought This Also Bought