Specifications
- Cap Size:33-400
- Cap Type:PE-Lined Black Phenolic Cap
- Capacity:1 L (33.8 oz.)
- Description:Amber Soda-Lime Safety Coated Bottle
- Cat. no.:16150-394
- Style:Boston Round
- Supplier No.:220946
Specifications
About this item
Create a safe work environment with Wheaton safety coated bottles.
Bottles with caps are used for distilled water, analytical standards and reagents. Metal foil liners are generally more resistant to organic solvents.
The strength, slip resistance and impact absorption qualities of the plastisol safety coating reduce potential breakage and exposure to hazardous materials. Wheaton '910' soda-lime flint amber glass protects light-sensitive samples.
If a bottle is accidentally broken, the PVC based plastisol coating on the outside of bottle retains the contents long enough for proper disposal. Ideal for lab or field applications, the bottle’s non-slip coating allows for greater ease and stability when handling wet or dry. The narrow Boston Round style is ideal when used for liquid. The bottles are manufactured from soda lime glass that conforms to USP Type III requirements.