Order Entry
ContactUsLinkComponent
Plastic Wide Mouth Laboratory Style Bottles, With Special Finish PP Linerless Caps
Lab Style Bottles, High-Density Polyethylene, Wide Mouth, Qorpak®
Catalog #: 16086-076
Supplier:  Qorpak
Small Business Enterprise
Plastic Wide Mouth Laboratory Style Bottles, With Special Finish PP Linerless Caps
Lab Style Bottles, High-Density Polyethylene, Wide Mouth, Qorpak®
Catalog #: 16086-076
Supplier:  Qorpak
Small Business Enterprise

Specifications

  • Cap Size:
    28 mm
  • Capacity:
    120 ml (4 oz.)
  • Cat. no.:
    16086-076
  • Supplier No.:
    PLA-03172

Specifications

About this item

Wide mouth HDPE lab style bottles are supplied with linerless polypropylene caps.

  • Ideal for high-speed filling, demanding pharmaceutical specifications, and the rough handling of interstate shipping

Excellent chemical resistance and the leak resistant closure make these ideal for storing, shipping, and packaging liquids for long periods. Linerless white polypropylene caps provide good impact strength, cost effectiveness, and pliability.

Packaging: Caps are attached on the convenience pack and enclosed on the bulk pack.
Customers Who Bought This Also Bought