Order Entry
ContactUsLinkComponent
NALGENE* Laboratory Bottles, Teflon* PFA, Narrow Mouth
Nalgene® Laboratory Bottles, Teflon® PFA, Narrow Mouth, Thermo Scientific
Catalog #: 16071-561
NALGENE* Laboratory Bottles, Teflon* PFA, Narrow Mouth
Nalgene® Laboratory Bottles, Teflon® PFA, Narrow Mouth, Thermo Scientific
Catalog #: 16071-561

Specifications

  • Cap Size:
    20-415
  • Capacity:
    30 ml (1 oz.)
  • Cat. no.:
    16071-561
  • Supplier No.:
    DS1630-0001

Specifications

About this item

Teflon® PFA resists temperature, chemicals, and corrosion.

  • Withstand temperatures from –270 to 250 °C (–454 to 482 °F)
  • Translucent bottles allow liquid levels to be seen
  • Linerless, leakproof closures made of Teflon PFA for a positive seal
  • Leakproof, Autoclavable

Note: Before autoclaving, set closure on top of bottle without engaging threads.

Inert to virtually all chemicals except molten alkali metals, fluorine at elevated temperatures, and certain complex halogenated compounds at elevated temperatures and pressures.

Customers Who Bought This Also Bought