Order Entry
ContactUsLinkComponent
Nalgene Packaging Bottles, HDPE, With Screw Caps, Thermo Scientific
Nalgene® Packaging Bottles, Wide Mouth, HDPE, with Closure, Bulk Packed, Thermo Scientific
Catalog #: 16059-604
Nalgene Packaging Bottles, HDPE, With Screw Caps, Thermo Scientific
Nalgene® Packaging Bottles, Wide Mouth, HDPE, with Closure, Bulk Packed, Thermo Scientific
Catalog #: 16059-604

Specifications

  • Cap Size:
    53-415
  • Capacity:
    500 ml (16.9 oz.)
  • Bottle Style:
    Wide mouth
  • Supplier No.:
    312189-0016
  • Height with Cap:
    168 mm

Specifications

About this item

These economical, break-resistant bottles are made from high-density polyethylene.

  • Available in natural or amber colors
  • Guaranteed leakproof

Natural sample bottles allow easy viewing of liquid level. Amber bottles reduce UV light transmission for protection of light-sensitive liquids.

Ordering information: Supplied with polypropylene screw caps.
Customers Who Bought This Also Bought