Order Entry
ContactUsLinkComponent
Nalgene Square Media Bottles, PETG, Sterile, Tray Pack Thermo Scientific
Nalgene® Square Media Bottles, PETG, Sterile, Thermo Scientific
Catalog #: 16059-494
Nalgene Square Media Bottles, PETG, Sterile, Tray Pack Thermo Scientific
Nalgene® Square Media Bottles, PETG, Sterile, Thermo Scientific
Catalog #: 16059-494

Specifications

  • Cap Size:
    38-430 mm
  • Capacity:
    125 ml (4.2 oz.)
  • Neck I.D.:
    28 mm
  • Outer Diameter:
    54 mm square
  • Supplier No.:
    342020-0125

Specifications

About this item

Nalgene® polyethylene terephthalate copolyester (PETG) square media bottles are an inexpensive alternative to glass media bottles.

  • Available with or without a natural HDPE screw cap closure
  • Bottles and closures are not autoclavable
  • Break-resistant, durable, and leakproof

Transparency and reduced permeability to CO₂/O₂ allow media storage for up to six months. Bottles and closures are radiation-sterilized and nonpyrogenic. Bottle 16159-999 (2 L size) features molded-in handgrips.

Ordering information: Packaged in shrink-wrapped trays. For additional screw cap/closure options, see 16060-026 series.
Customers Who Bought This Also Bought