Order Entry
ContactUsLinkComponent
VWR* Sample Bottles, High-Density Polyethylene, Narrow Mouth
VWR® Narrow Mouth Sample Bottles, HDPE
Catalog #: 16058-870
Supplier:  VWR International
Small Business Enterprise
VWR* Sample Bottles, High-Density Polyethylene, Narrow Mouth
VWR® Narrow Mouth Sample Bottles, HDPE
Catalog #: 16058-870
Supplier:  VWR International
Small Business Enterprise

Specifications

  • Cap Size:
    28-400
  • Capacity:
    946 ml (32 oz.)
  • Cat. no.:
    16058-870

Specifications

About this item

Lightweight, economy bottles offer an outstanding value for high volume sampling requirements.

  • Lightweight
  • Economical
  • Narrow-mouthed design
  • HDPE construction
  • Straight sides

Bottles have a straight-side commercial design. Tough polypropylene caps with polyethylene foam liners ensure tight seals.

Customers Who Bought This Also Bought